Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salicylic acid, Honeywell Fluka™

Catalog Number 60090386
Click to view available options
1KG POLY BOTTLE
69-72-7
338
CHEBI:16914
69-72-7
338
CHEBI:16914

Meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Appearance Complying
Percent Purity 99.5 to 100.5 %
Grade Reagent Pharmacopeia Acids
Quantity 1 kg
Loss on Drying Max. 0.3 %
Packaging Plastic Bottle
Physical Form Crystalline
Recommended Storage Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit