Learn More
ICU Medical Safe-T-Lance™ Safety Lancets
Safe-T-Lance™ Safety Lancets with retractable safety technology for capillary blood sampling
Supplier: ICU Medical 10081
Description
- Retractable safety technology for capillary blood sampling
- Automatic retraction reduces needlestick injuries and cross-contamination exposure
- Provides a quick, gentle puncture
- Needle is shielded before and after use
- Available in six different sizes for multiple types of applications
- Lancets are color coded for easy reference
Specifications
Built-in Control | |
18 ga | |
Yellow |
200/Box | |
High Flow | |
Yes |