Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ S-TEX 376SC Foam Nitrile Coated Cut Resistant Gloves

Catalog Number p-7192247
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Engineered with steel-reinforced Hagane Coil™ technology to provide ANSI A4 cut protection. Foam nitrile coating enhances grip capabilities, in addition to resisting hydrocarbons and oil.

  • Engineered with steel-reinforced Hagane Coil™ technology to provide ANSI A4 cut resistance
  • Lined with Kevlar™ aramid fibers for additional strength
  • Gauntlet style reinforced safety cuff extends cut protection up to the forearm
  • Nitrile coating is resistant to hydrocarbons, grease, and oil
  • Additional foam nitrile palm dip enhances wet and dry grip capabilities
  • Ergonomically designed hand mold reduces hand fatigue
ANSI Abrasion Rating ANSI 5
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Cuff Material Reinforced
Coating Material Nitrile Foam
Color Black, Blue
Cut Resistance Rating ANSI A4
Exterior Finish Partially Textured
Hand Specific
Material Polyester, Steel Fiber
Product Type Cut-resistant Gloves
Size Medium
EN388 Standard 4X41D
For Use With (Application) Drainage, HVAC, Mechanical Work, and Window Manufacturing
Glove Cuff Style Safety
Liner Material Aramid Fiber
Puncture Rating ANSI/ISEA 105-16 Level 3
Series S-TEX 376SC
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit