Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S-Tag Epitope Tag Antibody - BSA Free, Novus Biologicals™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600511
Description
S-Tag Epitope Tag Polyclonal antibody specifically detects S-Tag Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.Specifications
S-Tag Epitope Tag | |
Polyclonal | |
Unconjugated | |
PBS with 0.09% Sodium Azide | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:1000 - 1:30000, ELISA Primary-1:1000-1:30000; Coating: 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:400, Immunoprecipitation 1 - 4 ug/mg lysate | |
KETAAAKFERQHMDS, S 7 Epitope tag | |
Rabbits were immunized with S-Tag (KETAAAKFERQHMDS) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. | |
0.1 mL | |
Epitope Tags | |
Store at 4C. Do not freeze. | |
Epitope Tag |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction