Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laxco™ Rubber Grip for LMC-1000 Series Microscopes
Colored Hand Grips designed to be used with LMC-1000 Series Microscopes.
Specifications
For Use With (Equipment) | LMC-1000 Series Microscopes |
---|---|
Material | Rubber |
Warranty | 5 years optical and mechanical, 1 year electrical from date of purchase |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
LMCP0104792
|
Laxco™
LMCP0104792 |
Green | N/A | N/A | N/A | ||||
Description
- Choice any combination of colored handgrips for Laxco™ LMC-1000 series microscopes
- Color code them based off from their use, classroom, school colors, or just your favorite color
- Make your microscope unique, and more personalized
- Hand grips are make of a soft, no-slip rubber, with finger indents to make carrying of your microscope easy and comfortable
- Order them at the time of purchase of your microscope and we will install them before they ship, or add them to your existing microscope yourself
- NOTE: for use with Laxco LMC-1000 biological series microscopes ONLY
Specifications
LMC-1000 Series Microscopes | |
Rubber | |
5 years optical and mechanical, 1 year electrical from date of purchase |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title