Learn More
Chemglass Life Sciences Bump Trap, 500mL, Rotary Evaporator, Modified, 24/40 Top Outer, 24/40 Lower Inner
Supplier: Chemglass Life Sciences CG132109
Bump Trap, 500mL, Rotary Evaporator, Modified, 24/40 Top Outer, 24/40 Lower Inner. Similar to CG-1320, but with the addition of two 4mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask. Traps have a 24/40 top outer joint and the listed size of lower inner joint.