Learn More
Independent Forensics RSID™ Saliva Universal Buffer 25 tests/kit
Supplier: Independent Forensics 0130
The first specific test for Human α-Amylase. Animal Saliva does NOT cross-react with this test procedure. Detect as little as 1µL of human saliva. RSID™-Saliva results correlate with STR procedure. Strip test results are complete in 10 minutes.