Learn More
Diapharma Group, Inc. Rox Factor IX, 2 x 50 tests
Supplier: Diapharma Group, Inc. 900020
Rox Factor IX is a chromogenic assay kit based upon FXa generation for the determination of Factor IX (FIX) activity in FIX preparations, including potency assignment of FIX concentrates. For determination of Factor IX activity in plasma and Factor IX preparations, including potency assignment of FIX concentrates.