Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ mySPIN™ Mini Centrifuge Series Rotors
Spin 0.2mL and 1.5/2.0mL tubes with Thermo Scientific™ mySPIN™ Mini Centrifuge Series Rotors.
$56.30 - $99.30
Specifications
Material | Reinforced Glass Fiber |
---|---|
Type | Fixed Angle Rotor |
Biocontainment | No |
Catalog Number | Mfr. No. | Capacity | Max. RCF | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Max. RCF | For Use With (Equipment) | Price | Quantity & Availability | |||
75-004-062
|
Thermo Scientific™
75004062 |
6 x 1.5/2 mL | 2,000 x g | mySPIN 6 Mini Centrifuge |
Each for $56.30
|
|
|||
75-004-063
|
Thermo Scientific™
75004063 |
16 x 0.2 mL | 2,000 x g | mySPIN 6 Mini Centrifuge |
Each for $56.30
|
|
|||
75-004-084
|
Thermo Scientific™
75004084 |
32 x 0.2 mL | 9,800 x g | mySPIN 12 Mini Centrifuge |
Each for $99.30
|
|
|||
75-004-083
|
Thermo Scientific™
75004083 |
12 x 1.5/2 mL | 9,800 x g | mySPIN 12 Mini Centrifuge |
Each for $99.30
|
|
|||
Description
Spin 0.2mL and 1.5/2.0mL tubes with Thermo Scientific™ mySPIN™ Mini Centrifuge Series Rotors.
Specifications
Reinforced Glass Fiber | |
Fixed Angle Rotor | |
No |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title