Learn More
Pendotech Pendotech 1/4 Inch Hose Barb Single Use Rotary Flow Meter
Supplier: Pendotech FM22WV
Single Use Rotary Flow Meter, non-sterile, PVDF, 1/4 inch hose barb,0.1-2 lpm, clip mount. With individual calibrations.