Learn More
Cole-Parmer Stuart Roller Shaker, 6 Rollers, Variable Speed Digital, 120 VAC.
Supplier:  Cole-Parmer 0465305
Encompass_Preferred
Solidly constructed roller shakers are recommended for mixing samples, viscous substances and liquid-solid suspensions where minimum aeration is required. Shakers accommodate most tube and bottle sizes. The stacking system (sold separately) allows you to stack up to three shakers, providing expanded volume without taking valuable bench space.
Catalog No. NC1169090
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.