Learn More
OHAUS™ Rocking Incubating Shaker
OHAUS Rocking Incubating Shakers are designed to incubate samples from 5° above ambient to 65°C to provide accurate and repeatable results.
Supplier: OHAUS™ 30391980/EMD
Description
- Rocking shakers provide a see-saw like motion and feature LED displays with touchpad controls
- Electronic tilt adjustment from 0 to 15° while unit is operating
- LED displays for temperature, speed and time
- Timer with audible alarm
Specifications
0 to 15° | |
110 V | |
5°C to 65°C | |
3.74 A | |
50/60 Hz | |
27.4 cm | |
41.9 cm | |
10.1in. | |
25.7 cm | |
13.5 lb. | |
ISRK04HDG | |
3 wir cord and plug |
1 to 50 rpm | |
120 V | |
420 W | |
5° F to 149°F | |
10.8in. | |
10 in. | |
25.4 cm | |
7.5 in. | |
19.1 cm | |
6.1 kg | |
80% | |
1 sec. to 160 hr. |