Learn More
Chrono Log Corporation Ristocetin Cofactor Assay
SDP
Supplier:  Chrono Log Corporation 299
Encompass
Ristocetin Cofactor Assay is for use in the quantitative determination of Factor VIII Ristocetin Cofactor activity in citrated plasma.
Catalog No. NC0999310