Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversified Spaces™ Rinse Away Sink
Large sloping fiberglass sink with unique design to prevent splashes
Supplier: Diversified Spaces™ 3020KADA
Description
- Includes a hot and cold mixing faucet
- Limited Lifetime Warranty
- Made in the USA
- Overall dimensions: 80 in. W x 24 in. D x 38 in. H (34 in. H at the sink)
- Removable panel to access plumbing
- Sink is constructed of fiberglass to retain spilled materials and liquids
- The base consists of two oak storage cabinets with a drawer over the door with an adjustable shelf
- The counter is sloped with an integral sink, raised edges and coved corners
Warranty and Services
Limited Lifetime
Specifications
MAS Green Certified | |
Faucets with ADA Compliant Paddle Handles | |
24 x 80 x 38 in. | |
38 in. | |
234 lb. |
Oak, Fiberglass | |
4 | |
80 in. | |
24 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction