Learn More
Pyramex Safety Products Ridgeline™ Vented Full Brim Hard Hat
Ridgeline™ Vented Full Brim Hard Hat is strong, yet ultra-light for seemingly weightless protection.
$173.00 - $204.00
Specifications
Certifications/Compliance | Ansi Z89.1 |
---|---|
For Use With (Application) | Construction |
Product Type | 4-point Ratchet Suspension |
Size | 6.5 to 8 |
Suspension Type | 4-point ratchet suspension |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
19-555-101
|
Pyramex Safety Products
HP54110V |
White |
Case of 12 for $204.00
|
|
|||||
19-555-103
|
Pyramex Safety Products
HP54113V |
Gray |
Case of 12 for $204.00
|
|
|||||
19-555-106
|
Pyramex Safety Products
HP54130V |
Yellow |
Case of 12 for $173.00
|
|
|||||
19-555-107
|
Pyramex Safety Products
HP54160V |
Blue |
Case of 12 for $173.00
|
|
|||||
Description
- ABS material
- Ratchet suspension is easy to adjust and allows the wearer to modify the fit while wearing hard hat
- Soft brow pad is replaceable
- Replaceable suspensions and headbands also available
- Custom imprinting available on hard hats
- 4-point suspensions can be converted to a 6-point suspension with the HP6PTSUS
- Meets ANSI Z89.1-2014, Cert. Mod. - CAN/CSA Z94.1-15, Type 1, Class E, G, and C
Specifications
Ansi Z89.1 | |
4-point Ratchet Suspension | |
4-point ratchet suspension | |
Full Brim |
Construction | |
6.5 to 8 | |
Vented |