Learn More
U.S. Pharmacopeia Riboflavin, 83-88-5, MFCD00005022, 500mg
Supplier: U.S. Pharmacopeia 1603006
Molecular formula C17H20N4O6, Molecular weight 376.36 g/mol, Melting Point 527 - 554 °F (275 - 290 °C) (decomposes), Synonyms: Vitamin B2