Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Black Stallion™ Elkskin Leather Welder's Gloves with Nomex™ Lining
Click to view available options
Size:
Medium
Large
X-Large
Description
- Elkskin leather construction is extra thick to provide heat, flame, and abrasion resistance
- DuPont™ Nomex liner offers additional fire-resistant protection
- Aramid Fiber stitching for spark and abrasion resistance
- Reversed grain leather palm enhances grip, and provides a softer feel on the hand
- Thumb strap provides additional reinforcement or high stress areas
Specifications
Color | Gold |
Material | Elkskin Leather |
Size | Medium |
For Use With (Application) | Stick Welding |
Glove Cuff Color | White |
Glove Cuff Style | Gauntlet |
Liner Material | Nomex™ |
Palm Type | Grain Leather |
Series | 850 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction