Learn More
Cole-Parmer Reversible 16-Point Vertical Temperature Label, -3-13C/26-56F; 10/Pk
Supplier: Cole-Parmer 0903719
Digi-Sense Reversible 16-Point Vertical Temperature Label, -3-13C/26-56F; 10/Pk, Each style covers a wide range of temperatures, for use in many different applicationNon-toxic / non-hazardous temperature measuring alternativeEasy to read and unbreakable, These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.