Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RETSCH Grinding Containers for Knife Mill GRINDOMIX GM 200
For use with GM 200 GRINDOMIX Knife Mills.
Supplier: RETSCH 030450047
Description
- Suitable for soft and elastic materials and for all samples with high water, oil or fat content
- Can be sterilized and autoclaved to a limited extent
- Made of polypropylene
- Capacity: 1L
Specifications
Grinding Container | |
For soft and elastic materials and for all samples with high water, oil or fat content | |
Polypropylene |
1 L | |
GM 200 GRINDOMIX Knife Mills |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title