Learn More
Clontech Labs 3P Retro-X™ Concentrator, 500 ml
Supplier: Clontech Labs 3P 631456
The Retro-X Concentrator is a complete reagent for the concentration of infectious retroviral vector particles. This reagent provides a scalable alternative to ultracentrifugation for viral particle concentration. Vector supernatants can be concentrated 10–100 fold, depending upon the volumes and envelope used. 500ml