Learn More
Fine Science Tools RETRACTORS
Supplier: Fine Science Tools 1701211
Product: Weitlaner-Locktite Retractor/11cm/ Teeth: 2x3 Blunt/ Teeth Length: 16mm / Teeth Width: 12mm/ Maximum Spread: 4.5cm/ Alloy: Stainless Steel/ Length: 11cm/ Unit: Each