Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Mini pHPerch Storage Unit
Designed for pH meter supplies
Supplier: Restek™ 25147
Description
- Designed for pH meter supplies
- Small footprint, high storage capacity, easy access, built-in wiper dispenser
Specifications
Storage Unit | |
33 x 30.4 x 15.2 cm (13 x 12 x 6 in.) | |
Built-in wiper dispenser |
Small footprint, high storage capacity, easy access | |
pH meter supplies |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction