Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix Respiratory Syncytial Virus Type A Culture Fluid
Infectious virus preparation designed to meet research and product development needs
Supplier: ZeptoMetrix 0810040ACF
Description
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Specifications
ISO, 13485 | |
Solution | |
Respiratory Syncytial Virus Infectious Culture Fluid | |
1 mL |
-65°C | |
Vial | |
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays |
Safety and Handling
ShelfLife : 1 Year
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction