Learn More
Andwin Scientific RESERVOIR WITH LID
SDP
Supplier:  Andwin Scientific 89049026
Encompass_Preferred
Made with high clarity autoclavable polypropylene, the Reservoir Reagent is ideal for use with multi-well pipettors. This product comes with a lid that features a snap-seal closure. The reservoir has an overall volume capacity of 175 mL. It is sterile and graduated.
Catalog No. NC0620381