Learn More
Chemglass Life Sciences Reservoir Only, Airfree, Schlenk
Supplier: Chemglass Life Sciences AF0513A23
Reservoir Only, Airfree, Schlenk. Replacement part for AF0513A21. Complete bubbler consists of the lower reservoir, adjustable inner tube, compression cap and Viton o-ring.