Learn More
MilliporeSigma™ Replacement Parts for Stirred Cells
Suitable for use with Amicon™ stirred cell apparatus
Specifications
Includes | Cap assembly (1524901), stirrer assembly (15247) and base (210000) |
---|---|
Type | Replacement Part |
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
8003SCPKIT
|
MilliporeSigma™
8003SCPKIT |
Amicon 8003 Series Stirred Cell | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
8200SCPKIT
|
MilliporeSigma™
8200SCPKIT |
Amicon 8200 Series Stirred Cell | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Small Replacement Parts for 8000 Series Stirred Cells include 2 O-rings (Mfr. Nos. 2122S, 2118S, 2015S), tubing (Mfr. No. 4500202), tubing fitting assembly (Mfr. No. 15025AM, and membrane holder (Mfr. No. 2214801)
- Large Replacement Parts for Stirred Cells include one each: cap assembly, stirrer assembly and base
Specifications
Cap assembly (1524901), stirrer assembly (15247) and base (210000) | |
Replacement Part |