Learn More
United Scientific™ Replacement Filters for Pipette Filling Devices
SDPSCI_ED
Replacement Filters are compatible with many types of pipette filling devices. Available in a variety of pore sizes.
Supplier:  United Scientific™ MPF45PK/5
 View more versions of this product
Catalog No. S43745
  • $2.83 / Each of 1
    Save 90%
    Reg : $27.25
    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Description
Description
  • In-line membrane filters prevent aerosol contamination.
  • Prevents liquid from entering the equipment by accident.
  • Available in both 0.2 μm and 0.45 μm.
  • Each pack contains 5 filters.
Specifications
Specifications
Polypropylene
Non-sterile
5 Pk.
Disc
Product Suggestions
Product Suggestions