Learn More
Glas Col LLC Heating Mantle
SDP
Supplier:  Glas Col LLC 100D RX12712E
Encompass_Preferred
Replacement element for multi position combo mantle (100D RTX12712 six place low temperature). Easily changeable single poisiton. To 400 degrees C internal operating temperature. 100W 120V.
Catalog No. NC1833298