Learn More
Fisherbrand™ Replacement Deuterium Lamp For Shimadzu UV and Visible HPLC Detector
Fisherbrand™ Replacement Deuterium Lamp For Shimadzu UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
Catalog Number | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | For Use With (Equipment) | Price | Quantity & Availability | ||||||
14-500-725
|
Shimadzu SPD-10A, AVP, AV, AVVP, M1 UV and Visible HPLC Detector | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||||
14-500-727
|
Shimadzu LC2010 Long-life D2 Long-life UV and Visible HPLC Detector | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||||
Description
- Long life
- Drop-in OEM replacement
- Traceable by serial number
Compliance
WEEE and ROHS compliant
Specifications
Replacement Detector Lamp | |
Deuterium |