Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Phadebact™ Monoclonal GC Positive Control

Extracts from pure cultures of Neisseria gonorrhoeae.

Supplier:  Thermo Scientific™ R60815

Catalog No. R60815

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.

This item is not returnable. View return policy

Description

Description

Phadebact™ Monoclonal GC Positive Control are extracts from pure cultures of Neisseria gonorrhoeae.

  • Antigen level is adjusted to give a clear cut-off reaction against homologous reagents
  • For use with Monoclonal GC Test
  • 1mL/Vial
  • Bactus AB
Phadebact™ is a registered trademark of MKL Diagnostics AB.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Positive
N. gonorrhoeae
2 Vials/Pk.
Phadebact Monoclonal GC Positive Control
Phadebact Monoclonal GC Positive Control
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit