Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Regular Fetal Bovine Serum PROMO
FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
Supplier: Corning™ MT35010CVS23
Description
- Sera is aseptically collected and processed in a cGMP facility
- Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
- Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
- Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion
- Regular FBS is sourced from herds in USDA approved countries (ex: Mexico, Honduras, Costa Rica, Nicaragua, etc.)
Specifications
cGMP | |
-25° to -15°C | |
500 mL | |
Triple Filtered, 0.1μm | |
Regular | |
Sustains Cell Growth and Metabolism | |
<30 mg/dL | |
7.6 ±0.8 | |
USDA Inspected |
Amber Yellow to Red | |
Fetal Bovine Serum | |
Sterile | |
Bovine Fetus | |
≤20 EU/mL | |
No | |
315 ±35 mOsm/kg | |
30 to 50 g/L |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title