Learn More
Biocare Medical, LLC Barcoded Reagent Labels kit
Supplier: Biocare Medical, LLC NM129
Reagent Label Kit (NM129) includes one label ribbon (NM002) and two reagent label rolls (NM029). Each label roll has 1500 labels. Compatible with: intelliPATH FLX®, Nemesis (3600 and 7200) and ONCORE automated staining platforms.