Learn More
Ted Pella Inc REG SQ ARRAY 1.2UM HOLES 300M
Supplier: Ted Pella Inc 688300AU50
UltrAuFoil®, Regular Square Array of 1.2µm Diameter Holes – 1.3µm Separation, mounted on a 300M Au grid, package of 50 grids