Learn More
Durect Corporation REFLEX KIT 100
Supplier: Durect Corporation 0009972
The Reflex Kit 100 includes applier, remover, and 100 clips. The Reflex wound closure system is ideal for mice and young rats. It provides a fast and effective alternative to sutures for closing incisions.