Learn More
Reference Electrode for ion-selective electrodes, MilliporeSigma™ Supelco™
reference electrode with bridge, PTFE diaphragm, filled with 3 M KCl
Manufacturer:  MilliporeSigma Supelco 168111EAF
Catalog No. 11-100-4106
Add to cart
Description
Description
Specifications
Specifications
For ion-selective electrodes
1 Ea.
S7 connector
Ion Selective Electrode Standards