Learn More
MilliporeSigma™ Supelco™ Rectangular TLC Developing Tanks, Complete
TLC Developing Tanks come complete with the developing tank and lid.
Supplier: MilliporeSigma™ Supelco™ Z2041961EA
Description
TLC developing tanks are used to run TLC plates for separation. It is important to maintain saturated conditions during the analysis which can be achieved by using small tanks, filter paper liners and sufficient solvents. The tank can be equilibrated for 30 mins prior to running the plate to yield good results.Specifications
Glass | |
Developing tank and lid | |
6.8 cm |
TLC | |
17.5 cm | |
6.2 cm |