Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edvotek™ Reagents For Bacterial Transformation: Readypour Luria Broth
For rapid preparation of bacterial cultures for transformation.
$28.75 - $30.00
Specifications
Content And Storage | Room Temperature |
---|---|
For Use With (Application) | Rapid preparation of bacterial cultures for transformation |
Quantity | 170 mL |
Description
- 170 mL
Specifications
Room Temperature | |
Rapid preparation of bacterial cultures for transformation | |
170 mL |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title