Learn More
DWK Life Sciences Kimble™ RAY-SORB™ 5mm NMR Tubes
SDP
Kimble KIMAX™ High Grade NMR Tubes processed with proprietary RAY-SORB™ treatment. KIMAX-HQ NMR tubes are our highest quality NMR tubes and are 100% gauged for wall thickness, concentricity and camber specifications. They are ideal for sealing directly to vacuum manifolds, joints or valves.
Specifications
Product Type NMR Tube
View More Specs
Products 2
Catalog Number Mfr. No. Camber (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Camber (Metric) Price Quantity & Availability  
K8972419001
SDP
View Documents Promotion Details
DWK Life Sciences
8972419001
9 camber Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
K8972459001
SDP
View Documents Promotion Details
DWK Life Sciences
8972459001
3.8 µm Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. N/A
Description
Description
  • RAY-SORB protects your valuable samples from the harmful effect of visible and UV wavelengths, while keeping all the critical characteristics of your specific tube needs intact
  • Attached polyethylene caps
  • Sandblasted marking spot
  • OD 4.97 (+0.0000/-0.013) mm
  • ID 4.20 (+0.013/-0.0000) mm
  • Wall thickness 0.375 mm
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
NMR Tube