Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rat IgG1, FITC, Clone: HRPN, Negative Control, EMD Millpore™
Rat Monoclonal Antibody
Supplier: MilliporeSigma™ MABF1780
Description
Specifically detects IgG1 Clone: HRPN in samples, and it is validated for Flow Cytometry
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.Specifications
IgG1 | |
Monoclonal | |
FITC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
Flow Cytometry | |
HRPN | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. This Monoclonal was purified from tissue culture supernatant via affinity chromatography. | |
Proprietary immunogen | |
100 μg | |
Inflammation, Immunology | |
Rat IgG1 | |
IgG1 |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
For Research Use Only
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'