Learn More
R&D Systems™ EvenCoat Goat Anti-Mouse IgG Coated Plates
used as a solid support for most sandwich ELISAs utilizing a mouse IgG capture antibody and a non-mouse IgG detection antibody, other applications include competitive ELISA, IgG isotyping, and hybridoma screening/selection.
$204.50 - $560.00
Specifications
Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
---|---|
Surface Treatment | EvenCoat Goat Anti-Mouse IgG |
For Use With (Application) | ELISA |
Specifications
Store the unopened product at 2 - 8° C. Do not use past expiration date. | |
EvenCoat Goat Anti-Mouse IgG | |
ELISA |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.