Learn More
MilliporeSigma™ Supelco™ radiello™ Diffusive Bodies
Designed to house radiello™ cartridge adsorbents during sampling
Supplier: MilliporeSigma™ Supelco™ RAD1202
Description
Diffusive bodies are threaded at one end for easy attachment to the radiello™ triangular support plate. Unlike most of the cartridge adsorbents, the diffusive bodies are reusable and can be cleaned with a mild detergent between use. The diffusive bodies will collect dust (especially during outdoor sampling) and replacement is recommended after 4-5 uses. Dimensions are 60 mm height x 16 mm diameter. There are four different diffusive bodies available. Each of which are application specific in design.Specifications
Yellow Polyethylene Body | |
Reduced Sampling Rates | |
20/Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
radiello is a trademark of Institi Clinici Scientifici Maugeri