Learn More
Se International Inc RADIATION ALERT MONITOR 4
Supplier: Se International Inc MONITOR4
Monitor 4, Size: 145 x 72 x 38mm, Gamma Sensitivity: 1000 cpm/mR/hr, Operating Range: 0 to 0.5, 0 to 5, 0 to 50 mR/hr. and 0 to 500, 0 to 5,000, 0 to 50,000 cpm, Temperature Range: -20degree to +55 deg.C