Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ RadHalo Spectroscopic Area Monitor Radio Communication Options
For use with RadHalo Spectroscopic Area Monitor models with the radio option
$242.00
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
19-175-826
|
Thermo Scientific™
COMC004162 |
Matrix Base Radio and Matrix Radio Repeater Pole Mount Kit |
Each for $242.00
|
|
|||||
19-175-824
|
Thermo Scientific™
MTXBR24L |
Matrix Base Radio - Single Laird 2.4GHz radio in IP67 rated housing | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
19-175-825
|
Thermo Scientific™
MTXR24L |
Matrix Radio Repeater - Dual 2.4 GHz Laird Radios | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Matrix Base Radio and Matrix Radio Repeater- Include rugged IP67 housing, 2.4 GHz radio, external antenna connection, power connection, ethernet connection and wall mount
- Require an appropriate 2.4 GHz antenna kit
- Do not include a pole mounting kit or screws/anchors for wall mount
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title