Learn More
Fisherbrand™ Racer Series Qualifier Eyewear
Click to view available options
Frame Color:
Black
Red
Lens Tint:
Clear
Dark Gray
Description
- Scratch-resistant, clear polycarbonate lenses block >99.9% of harmful UV radiation
- Wraparound design offers superior side coverage
- Lightweight frames are durable and comfortable
- Flared single-lens design provides sleek protection
- A soft nose bridge and soft, bayonet-style side arms grip the face and temples lightly but firmly for a comfortable fit
- Choose from black or red nylon frames
Specifications
Lens Tint | Clear |
Frame Color | Black |
Lens Material | Polycarbonate |
Frame Material | Nylon |