Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Rabbit Serum, Heat Inactivated
Rabbit Serum is used in various research applications and obtained from healthy laboratory animals
Supplier: Novus Biologicals™ S15110HNOV
Specifications
Store at -20°C in the dark. Avoid freeze-thaw cycles. | |
Rabbit | |
100 mL |
Rabbit Serum, Heat Inactivated | |
Rabbit Serum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title