Learn More
Rockland Immunochemicals Rabbit anti-Sheep Cell Hemolysin - C220-0002
Supplier: Rockland Immunochemicals C2200002
Sheep red blood cells coated with Rabbit anti-Sheep Cell Hemolysin have traditionally been used to assay the activity of the complement classical pathway in serum samples.