Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Antigen Retrieval Reagent Sampler
Contains three antigen retrieval reagents in a single pack
Supplier: R&D Systems™ CTS016
This item is not returnable.
View return policy
Description
- Use a heat-induced recovery of cell and tissue antigens
- Includes Antigen Retrieval Reagent-Basic (Mfr. No. CTS013), Antigen Retrieval Reagent-Acidic (Mfr. No. CTS014) and Antigen Retrieval Reagent-Universal (Mfr. No. CTS015) in 10X concentration
Antigen Retrieval Reagent-Basic and Antigen Retrieval Reagent-Acidic may change tissue morphology.
Applications:
Immunohistochemistry
Specifications
Store at Room temperature | |
Antigen Retrieval Reagent Sampler | |
Antigen retrieval |
10 X | |
50 mL each reagent | |
RUO |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title