Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Antigen Retrieval Reagent, Acidic
An antigen retrieval system using heat-induced recovery of cell and tissue antigens
Supplier: R&D Systems™ CTS014
This item is not returnable.
View return policy
Description
- Supplied in 10X concentration
May change tissue morphology.
Applications:
Immunohistochemistry
Specifications
Store at Room temperature | |
Antigen Retrieval Reagent-Acidic | |
50 mL | |
Antigen retrieval |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title