Learn More
CTL Scientific Supply Corp QUANTOFIX GLUCOSE,100 TESTS
SDP
Supplier:  CTL Scientific Supply Corp 91348
Encompass_Preferred
QUANTOFIX GLUCOSE are test sticks for the semi-quantitative determination of glucose content. Gradation 0 - 5 - 100 - 250 - 500 - 1000 - 2000 mg/l glucose. Contains 100 test strips
Catalog No. 50-117-3324
Due to product restrictions, please to purchase or view availability for this product.