Learn More
CTL Scientific Supply Corp QUANT. PERACETIC ACID,100STR
Supplier: CTL Scientific Supply Corp 91341
QUANTOFIX PERACETIC ACID 500 are test sticks for the semi-quantitative determination of peracetic acid. Gradation 50 - 100 - 200 - 300 - 400 - 500 mg/l. Contains 100 test strips